Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sri-48
Entrez Gene ID
191919
Gene Name
Serpentine Receptor, class I
Gene Aliases
CELE_ZC239.9
Chromosome Location
Chr.II: 3199583 - 3201342 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062069.3 NP_494470.1 5 - 6 709 114

View Full Product Details