Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-71
Entrez Gene ID
173695
Gene Name
Col_cuticle_N domain-containing protein
Gene Aliases
CELE_Y49F6B.10
Chromosome Location
Chr.II: 3500077 - 3502570 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062161.5 NP_494562.1 1 - 2 218 85

View Full Product Details