Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-97
Entrez Gene ID
184672
Gene Name
7TM_GPCR_Srx domain-containing protein
Gene Aliases
CELE_F19B10.7
Chromosome Location
Chr.II: 3667136 - 3671068 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062208.3 NP_494609.1 10 - 11 1128 97

View Full Product Details