Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sdz-12
Entrez Gene ID
184388
Gene Name
Zinc finger protein sdz-12
Gene Aliases
CELE_F12E12.5
Chromosome Location
Chr.II: 3730152 - 3731823 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062233.2 NP_494634.1 2 - 3 453 79

View Full Product Details