Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sea-2
Entrez Gene ID
173738
Gene Name
protein-coding
Gene Aliases
CELE_K10G6.3
Chromosome Location
Chr.II: 3950377 - 3972140 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306544.2 NP_001293473.1 7 - 8 5196 67
NM_001313639.3 NP_001300568.1 6 - 7 4956 67
NM_001313640.3 NP_001300569.1 5 - 6 4521 67
NM_001313642.3 NP_001300571.1 4 - 5 4035 67
NM_001313643.3 NP_001300572.1 5 - 6 4470 67
NM_001313644.3 NP_001300573.1 2 - 3 2883 67
NM_001377772.3 NP_001364836.1 10 - 11 5484 67
NM_001377773.3 NP_001364838.1 9 - 10 5049 67

View Full Product Details