Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
atg-9
Entrez Gene ID
178561
Gene Name
protein-coding
Gene Aliases
CELE_T22H9.2
Chromosome Location
Chr.V: 349385 - 359658 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070777.5 NP_503178.2 5 - 6 863 157

View Full Product Details