Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-65
Entrez Gene ID
353476
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_T05C3.8
Chromosome Location
Chr.V: 5483010 - 5484966 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182424.2 NP_872224.1 4 - 5 830 120

View Full Product Details