Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-29
Entrez Gene ID
353465
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_T09D3.6
Chromosome Location
Chr.V: 5337184 - 5339013 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182426.1 NP_872226.1 3 - 4 875 95

View Full Product Details