Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-38
Entrez Gene ID
353467
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_T19C4.8
Chromosome Location
Chr.V: 11152333 - 11155653 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182428.2 NP_872228.1 2 - 3 500 96

View Full Product Details