Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mlt-7
Entrez Gene ID
173775
Gene Name
Peroxidase mlt-7 light chain
Gene Aliases
CELE_ZK430.8
Chromosome Location
Chr.II: 4392672 - 4404850 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062376.6 NP_494777.1 5 - 6 1136 110

View Full Product Details