Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dnj-9
Entrez Gene ID
3565947
Gene Name
J domain-containing protein
Gene Aliases
CELE_F11G11.7
Chromosome Location
Chr.II: 4852056 - 4857880 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062471.5 NP_494872.2 2 - 3 633 80

View Full Product Details