Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cblc-1
Entrez Gene ID
3565644
Gene Name
MMACHC-like protein
Gene Aliases
CELE_ZK546.17
Chromosome Location
Chr.II: 4938595 - 4942870 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027353.7 NP_001022524.1 2 - 3 322 99

View Full Product Details