Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tbx-35
Entrez Gene ID
191239
Gene Name
Putative T-box protein 35
Gene Aliases
CELE_ZK177.10
Chromosome Location
Chr.II: 5524101 - 5525397 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062658.3 NP_495059.2 1 - 2 268 157

View Full Product Details