Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- tbc-2
- Entrez Gene ID
-
173986
- Gene Name
- TBC (Tre-2/Bub2/Cdc16) domain family
- Gene Aliases
-
CELE_ZK1248.10
- Chromosome Location
-
Chr.II: 5823856 - 5827443 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_062755.9 |
NP_495156.1 |
9 - 10 |
2579 |
81 |
- Gene Symbol
- ZK1248.11
- Entrez Gene ID
-
173987
- Gene Name
- SP-RING-type domain-containing protein
- Gene Aliases
-
CELE_ZK1248.11
- Chromosome Location
-
Chr.II: 5823853 - 5828588 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383850.1 |
|
12 - 13 |
3179 |
81 |
View Full Product Details