Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lec-5
Entrez Gene ID
173992
Gene Name
Galectin
Gene Aliases
CELE_ZK1248.16
Chromosome Location
Chr.II: 5839637 - 5841187 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062762.10 NP_495163.1 5 - 6 659 141
GenBank mRNA AB038506.1 - 4 - 5 632 141

View Full Product Details