Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fbf-2
Entrez Gene ID
174017
Gene Name
Fem-3 mRNA-binding factor 2
Gene Aliases
CELE_F21H12.5
Chromosome Location
Chr.II: 6078309 - 6091480 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381445.1 NP_001367592.1 1 - 2 154 78

View Full Product Details