Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sma-6
Entrez Gene ID
174044
Gene Name
Serine/threonine-protein kinase sma-6
Gene Aliases
CELE_C32D5.2
Chromosome Location
Chr.II: 6324458 - 6327355 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062870.7 NP_495271.1 8 - 9 986 155
GenBank mRNA AF104017.1 - 8 - 9 967 155

View Full Product Details