Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wrn-1
Entrez Gene ID
174081
Gene Name
Probable Werner syndrome ATP-dependent helicase homolog 1
Gene Aliases
CELE_F18C5.2
Chromosome Location
Chr.II: 6554977 - 6559310 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_062923.7 NP_495324.2 1 - 2 122 87

View Full Product Details