Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
glc-4
Entrez Gene ID
182971
Gene Name
Glutamate-gated ChLoride channel
Gene Aliases
CELE_C27H5.8
Chromosome Location
Chr.II: 7172204 - 7174936 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063088.5 NP_495489.2 3 - 4 698 109

View Full Product Details