Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pdl-1
Entrez Gene ID
191741
Gene Name
Phosphodiesterase delta-like protein
Gene Aliases
CELE_C27H5.1
Chromosome Location
Chr.II: 7183801 - 7185294 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063089.3 NP_495490.1 3 - 4 424 88

View Full Product Details