Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- dhs-7
- Entrez Gene ID
-
174183
- Gene Name
- DeHydrogenases, Short chain
- Gene Aliases
-
CELE_E04F6.7
- Chromosome Location
-
Chr.II: 7206990 - 7208460 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063099.5 |
NP_495500.1 |
3 - 4 |
702 |
126 |
- Gene Symbol
- E04F6.15
- Entrez Gene ID
-
184041
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_E04F6.15
- Chromosome Location
-
Chr.II: 7207001 - 7210287 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063100.3 |
NP_495501.1 |
6 - 7 |
2083 |
126 |
View Full Product Details