Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-55
Entrez Gene ID
191817
Gene Name
Serpentine receptor class delta-55
Gene Aliases
CELE_K02A2.2
Chromosome Location
Chr.II: 7408492 - 7410759 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063152.2 NP_495553.1 2 - 3 313 123

View Full Product Details