Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kcc-3
Entrez Gene ID
174212
Gene Name
protein-coding
Gene Aliases
CELE_K02A2.3
Chromosome Location
Chr.II: 7425939 - 7434282 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001351919.3 NP_001338828.1 4 - 5 296 137
NM_001351920.3 NP_001338797.1 3 - 4 283 137

View Full Product Details