Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ifg-1
Entrez Gene ID
174322
Gene Name
protein-coding
Gene Aliases
CELE_M110.4
Chromosome Location
Chr.II: 8216310 - 8222396 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027088.7 NP_001022259.1 9 - 10 3462 91
NM_001027089.7 NP_001022260.1 8 - 9 3330 91
NM_001373755.2 NP_001359723.1 8 - 9 3349 91
NM_001381521.1 NP_001368728.1 8 - 9 3323 91
NM_001383902.1 9 - 10 3423 91

View Full Product Details