Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-73
Entrez Gene ID
174366
Gene Name
protein-coding
Gene Aliases
CELE_W01C9.3
Chromosome Location
Chr.II: 8540712 - 8545393 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267229.4 NP_001254158.1 7 - 8 924 92
NM_001267230.4 NP_001254159.1 6 - 7 833 92

View Full Product Details