Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rol-6
Entrez Gene ID
174397
Gene Name
Cuticle collagen rol-6
Gene Aliases
CELE_T01B7.7
Chromosome Location
Chr.II: 8733050 - 8734239 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063457.4 NP_495858.1 1 - 2 138 82

View Full Product Details