Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rha-1
Entrez Gene ID
174417
Gene Name
protein-coding
Gene Aliases
CELE_T07D4.3
Chromosome Location
Chr.II: 8878496 - 8883659 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373779.1 NP_001360473.1 5 - 6 1047 104
NM_063489.6 NP_495890.2 5 - 6 1047 104

View Full Product Details