Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sym-2
Entrez Gene ID
174461
Gene Name
protein-coding
Gene Aliases
CELE_ZK1067.6
Chromosome Location
Chr.II: 9178514 - 9192877 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001313679.3 NP_001300608.1 1 - 2 155 153
NM_063559.9 NP_495960.2 11 - 12 2003 153
GenBank mRNA AY220985.1 - 5 - 6 1007 153

Gene Symbol
twk-4
Entrez Gene ID
192079
Gene Name
TWiK family of potassium channels
Gene Aliases
CELE_ZK1067.5
Chromosome Location
Chr.II: 9178514 - 9194433 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001356885.2 NP_001343566.1 - 2310 153

View Full Product Details