Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- sym-2
- Entrez Gene ID
-
174461
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_ZK1067.6
- Chromosome Location
-
Chr.II: 9178514 - 9192877 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_063559.9 |
NP_495960.2 |
3 - 4 |
486 |
118 |
- Gene Symbol
- twk-4
- Entrez Gene ID
-
192079
- Gene Name
- TWiK family of potassium channels
- Gene Aliases
-
CELE_ZK1067.5
- Chromosome Location
-
Chr.II: 9178514 - 9194433 on Build WBcel235
View Full Product Details