Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
exc-7
Entrez Gene ID
174506
Gene Name
Uncharacterized protein
Gene Aliases
CELE_F35H8.5
Chromosome Location
Chr.II: 9575971 - 9581560 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063656.5 NP_496057.1 3 - 4 682 127

View Full Product Details