Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-13A10
Entrez Gene ID
174522
Gene Name
Putative cytochrome P450 CYP13A10
Gene Aliases
CELE_ZK1320.4
Chromosome Location
Chr.II: 9660839 - 9663007 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063684.5 NP_496085.3 5 - 6 1348 70

View Full Product Details