Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-13A1
Entrez Gene ID
188361
Gene Name
Putative cytochrome P450 CYP13A1
Gene Aliases
CELE_T10B9.8
Chromosome Location
Chr.II: 9779181 - 9781997 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383923.1 1 - 2 98 75

View Full Product Details