Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pho-11
Entrez Gene ID
174547
Gene Name
Putative acid phosphatase 11
Gene Aliases
CELE_C05C10.4
Chromosome Location
Chr.II: 9931845 - 9934427 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063742.8 NP_496143.1 4 - 5 483 67

View Full Product Details