Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lec-3
Entrez Gene ID
174557
Gene Name
protein-coding
Gene Aliases
CELE_ZK892.1
Chromosome Location
Chr.II: 9985007 - 10000351 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027360.5 NP_001022531.1 9 - 10 929 78
NM_001027361.4 NP_001022532.1 10 - 11 1139 78
NM_001027362.7 NP_001022533.1 10 - 11 1086 78
NM_001276739.3 NP_001263668.1 10 - 11 877 78
NM_063758.8 NP_496159.1 10 - 11 912 78
GenBank mRNA AB038504.1 - 9 - 10 819 78

View Full Product Details