Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bath-38
Entrez Gene ID
174567
Gene Name
BTB and MATH domain-containing protein 38
Gene Aliases
CELE_F52H3.3
Chromosome Location
Chr.II: 10042780 - 10045270 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063771.5 NP_496172.1 2 - 3 318 95

View Full Product Details