Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
daf-19
Entrez Gene ID
174577
Gene Name
protein-coding
Gene Aliases
CELE_F33H1.1
Chromosome Location
Chr.II: 10157520 - 10169354 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026968.4 NP_001022139.1 8 - 9 1146 78
NM_001026969.4 NP_001022140.1 9 - 10 1220 78
NM_001267307.4 NP_001254236.1 9 - 10 1233 78
NM_001267308.3 NP_001254237.1 5 - 6 813 78
NM_001306556.3 NP_001293485.1 5 - 6 695 78
GenBank mRNA AF226156.1 - 9 - 10 1243 78
AF233652.1 - 8 - 9 1168 78
EU812221.1 - 5 - 6 813 78

View Full Product Details