Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tnc-2
Entrez Gene ID
174612
Gene Name
Troponin C, isoform 2
Gene Aliases
CELE_ZK673.7
Chromosome Location
Chr.II: 10466852 - 10468358 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063850.6 NP_496251.1 4 - 5 409 71

View Full Product Details