Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-61
Entrez Gene ID
174613
Gene Name
C-type LECtin
Gene Aliases
CELE_ZK666.7
Chromosome Location
Chr.II: 10488730 - 10490537 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063859.5 NP_496260.1 4 - 5 554 128

View Full Product Details