Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pro-1
Entrez Gene ID
174622
Gene Name
Pre-rRNA-processing protein pro-1
Gene Aliases
CELE_R166.4
Chromosome Location
Chr.II: 10539436 - 10541996 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063870.6 NP_496271.1 3 - 4 988 105

View Full Product Details