Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gbh-1
Entrez Gene ID
174648
Gene Name
Probable gamma-butyrobetaine dioxygenase
Gene Aliases
CELE_D2089.5
Chromosome Location
Chr.II: 10690452 - 10693863 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063904.7 NP_496305.1 4 - 5 778 80

View Full Product Details