Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bli-1
Entrez Gene ID
174653
Gene Name
Cuticle collagen bli-1
Gene Aliases
CELE_C09G5.6
Chromosome Location
Chr.II: 10708923 - 10712156 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063910.7 NP_496311.2 1 - 2 232 84

View Full Product Details