Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lir-3
Entrez Gene ID
174707
Gene Name
Transcription factor lir-3
Gene Aliases
CELE_F37H8.1
Chromosome Location
Chr.II: 11183047 - 11184807 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_063994.6 NP_496395.1 1 - 2 70 63

View Full Product Details