Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cut-1
Entrez Gene ID
174720
Gene Name
Cuticlin-1
Gene Aliases
CELE_C47G2.1
Chromosome Location
Chr.II: 11269122 - 11271660 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064009.4 NP_496410.1 3 - 4 1088 109

View Full Product Details