Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rsp-2
Entrez Gene ID
174747
Gene Name
Probable splicing factor, arginine/serine-rich 2
Gene Aliases
CELE_W02B12.2
Chromosome Location
Chr.II: 11451441 - 11452955 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064040.7 NP_496441.1 3 - 4 462 58

View Full Product Details