Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rga-1
Entrez Gene ID
174751
Gene Name
protein-coding
Gene Aliases
CELE_W02B12.8
Chromosome Location
Chr.II: 11464894 - 11468653 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027219.6 NP_001022390.1 1 - 2 130 62

View Full Product Details