Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pcs-1
Entrez Gene ID
174775
Gene Name
protein-coding
Gene Aliases
CELE_F54D5.1
Chromosome Location
Chr.II: 11574941 - 11577740 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129143.5 NP_001122615.1 3 - 4 801 72
NM_001129144.4 NP_001122616.1 2 - 3 295 72

View Full Product Details