Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nep-1
Entrez Gene ID
174787
Gene Name
Neprilysin-1
Gene Aliases
CELE_ZK20.6
Chromosome Location
Chr.II: 11654214 - 11657172 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064089.6 NP_496490.2 3 - 4 2172 69

View Full Product Details