Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-27
Entrez Gene ID
189224
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_W05H5.4
Chromosome Location
Chr.II: 12448541 - 12449951 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064246.2 NP_496647.1 1 - 2 206 130

View Full Product Details