Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-68
Entrez Gene ID
188674
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_T21B4.5
Chromosome Location
Chr.II: 12507338 - 12508452 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_064267.1 NP_496668.1 1 - 2 350 90

View Full Product Details