Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tbc-14
Entrez Gene ID
174986
Gene Name
protein-coding
Gene Aliases
CELE_E01G4.1
Chromosome Location
Chr.II: 13436923 - 13447593 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001330963.2 NP_001317775.1 3 - 4 641 85
NM_001330964.4 NP_001317776.1 3 - 4 647 85

View Full Product Details